Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM6/CD66c Rabbit anti-Human, Clone: 2, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | CEACAM6/CD66c |
---|---|
Clone | 2 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
CEACAM6/CD66c Monoclonal specifically detects CEACAM6/CD66c in Human samples. It is validated for ELISA.Specifications
CEACAM6/CD66c | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen | |
Recombinant Human CEACAM6/CD66c Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
2 | |
ELISA | |
Unconjugated | |
Rabbit | |
Cancer | |
0.2 um filtered solution in PBS | |
4680 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title