Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM7 Antibody (962720), PE, Novus Biologicals™
SDP

Catalog No. NB370026 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370026 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370026 Supplier Novus Biologicals Supplier No. FAB44782P

Mouse Monoclonal Antibody

CEACAM7 Monoclonal antibody specifically detects CEACAM7 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, CyTOF

Specifications

Antigen CEACAM7
Applications Flow Cytometry, ELISA, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 962720
Conjugate PE
Dilution Flow Cytometry, ELISA, Immunohistochemistry, CyTOF-ready
Formulation PBS
Gene Alias Carcinoembryonic antigen CGM 2, Carcinoembryonic antigen CGM2, Carcinoembryonic antigen gene family member 2, Carcinoembryonic antigen related cell adhesion molecule 7, CEA, CEACAM 7, CGM 2, CGM2
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived recombinant human CEACAM-7, Thr36-Asn233, Accession # Q14002
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1087
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.