Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM8/CD66b Antibody (913542), HRP, Novus Biologicals™
SDP

Catalog No. NB370506 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370506 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370506 Supplier Novus Biologicals Supplier No. FAB4246H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEACAM8/CD66b Monoclonal specifically detects CEACAM8/CD66b in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen CEACAM8/CD66b
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 913542
Conjugate HRP
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95
Host Species Mouse
Immunogen E.coli-derived recombinant human CEACAM-8/CD66b, Gln35-His141, Accession # P31997
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1088
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.