Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM8/CD66b Antibody (G10F5) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013737 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013737 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013737 Supplier Novus Biologicals Supplier No. NBP280664
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEACAM8/CD66b Monoclonal specifically detects CEACAM8/CD66b in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.

Specifications

Antigen CEACAM8/CD66b
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
Classification Monoclonal
Clone G10F5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 1ug/million cells, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen : 1:10-1:500, CyTOF-ready
Formulation PBS
Gene Alias Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95
Gene Symbols CEACAM8
Host Species Mouse
Immunogen Human peripheral blood cells.
Purification Method IgM purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1088
Test Specificity Predicited to cross react with Chimpanzee.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.