Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM8/CD66b Antibody (G10F5), Novus Biologicals™
SDP

Catalog No. NB10077808 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10077808 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10077808 Supplier Novus Biologicals Supplier No. NB10077808
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 4 publications

CEACAM8/CD66b Monoclonal specifically detects CEACAM8/CD66b in Human, Mouse samples. It is validated for CyTOF, Flow Cytometry, IF/IHC, Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).

Specifications

Antigen CEACAM8/CD66b
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone G10F5
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 33122293), Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry, CyTOF-ready
Gene Accession No. P31997
Gene Alias Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, cd66b clone g10f5, cd66b G10F5, cd66b granulocyte marker, cd66b ihc, cd66b marker, cd66b neutrophil marker, cd66b neutrophils, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95
Gene Symbols CEACAM8
Host Species Mouse
Immunogen Human peripheral blood cells.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1088
Test Specificity CEACAM8 (G10F5). Expected Cross-Reactivity based on immunogen sequence: Chimpanzee.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.