Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM8/CD66b Antibody (SR2195), Novus Biologicals™
SDP

Catalog No. NB387935 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387935 50 μL
NB258574 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB387935 Supplier Novus Biologicals Supplier No. NBP32196550UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CEACAM8/CD66b Monoclonal antibody specifically detects CEACAM8/CD66b in Human samples. It is validated for Western Blot

Specifications

Antigen CEACAM8/CD66b
Classification Monoclonal
Clone SR2195
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95
Host Species Rabbit
Immunogen A synthesized peptide derived from human CEACAM8/CD66b (Uniprot number: P31997)
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1088
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.