Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEBP Beta Antibody (47A1), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP104266FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104266FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104266FR Supplier Novus Biologicals Supplier No. NBP104266FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEBP Beta Monoclonal specifically detects CEBP Beta in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CEBP Beta
Applications Western Blot, ELISA
Classification Monoclonal
Clone 47A1
Conjugate DyLight 680
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C/EBP beta, C/EBP-beta, CCAAT/enhancer binding protein (C/EBP), beta, CCAAT/enhancer-binding protein beta, CRP2, IL6DBP, interleukin 6-dependent DNA-binding protein, LAPTCF-5, Liver activator protein, liver-enriched transcriptional activator protein, MGC32080, NF-IL6, Nuclear factor NF-IL6, nuclear factor of interleukin 6, TCF5NFIL6, Transcription factor 5
Gene Symbols CEBPB
Host Species Mouse
Immunogen Recombinant human C/EBP-beta (1-271 aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1051
Test Specificity Anti-human C/EBP beta mAb, clone 47A1, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human C/EBP beta protein.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.