Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEBP Delta Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB396462 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396462 0.025 mg
NBP241192 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396462 Supplier Novus Biologicals Supplier No. NBP2411920.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CEBP Delta Polyclonal specifically detects CEBP Delta in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CEBP Delta
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias C/EBP-delta, CCAAT/enhancer binding protein (C/EBP), delta, CCAAT/enhancer-binding protein delta, CELF, CRP3, NF-IL6-betaC/EBP delta, Nuclear factor NF-IL6-beta
Gene Symbols CEBPD
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human CEBPD. The immunogen is located within amino acids 170 - 220 of CEBPD. Amino Acid Squence: CKRGPDRGSPEYRQRRERN
Molecular Weight of Antigen 30 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1052
Test Specificity Multiple transcript variants encoding different isoforms have been found for this gene.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.