Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEBP Delta Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB120739 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120739 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120739 Supplier Novus Biologicals Supplier No. NB11085519C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CEBP Delta Polyclonal antibody specifically detects CEBP Delta in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, KnockDown

Specifications

Antigen CEBP Delta
Applications Western Blot, ELISA, Immunohistochemistry, KnockDown
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, ELISA, Immunohistochemistry, Knockdown Validated
Formulation 50mM Sodium Borate
Gene Alias C/EBP-delta, CCAAT/enhancer binding protein (C/EBP), delta, CCAAT/enhancer-binding protein delta, CELF, CRP3, NF-IL6-betaC/EBP delta, Nuclear factor NF-IL6-beta
Host Species Rabbit
Immunogen This affinity purified CEBP Delta Antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant protein corresponding to the amino terminus of mouse CEBP Delta. (Uniprot: Q00322)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1052
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.