Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEBP Delta Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB199419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB199419 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB199419 Supplier Novus Biologicals Supplier No. NBP241192IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CEBP Delta Polyclonal antibody specifically detects CEBP Delta in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen CEBP Delta
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias C/EBP-delta, CCAAT/enhancer binding protein (C/EBP), delta, CCAAT/enhancer-binding protein delta, CELF, CRP3, NF-IL6-betaC/EBP delta, Nuclear factor NF-IL6-beta
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human CEBPD. The immunogen is located within amino acids 170 - 220 of CEBPD. Amino Acid Squence: CKRGPDRGSPEYRQRRERN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Endocrinology, Immunology, Innate Immunity, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1052
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.