Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEBP Delta Antibody, Novus Biologicals™
SDP

Catalog No. NBP221568 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221568 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221568 Supplier Novus Biologicals Supplier No. NBP221568
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CEBP Delta Polyclonal antibody specifically detects CEBP Delta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, ChIP assay.

Specifications

Antigen CEBP Delta
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.93mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (ChIP)
Formulation PBS pH7, 20% Glycerol with 0.025% Proclin 300
Gene Accession No. P49716
Gene Alias C/EBP-delta, CCAAT/enhancer binding protein (C/EBP), delta, CCAAT/enhancer-binding protein delta, CELF, CRP3, NF-IL6-betaC/EBP delta, Nuclear factor NF-IL6-beta
Gene Symbols CEBPD
Host Species Rabbit
Immunogen Full length human C/EBP delta Recombinant protein.
Molecular Weight of Antigen 28 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1052
Test Specificity Human, Mouse. Expected to cross react based on sequence identity: Mouse (85%), Rat (86%), Bovine (85%) Expected to cross react based on sequence identity: Mouse (85%), Rat (86%), Bovine (85%).
Target Species Rat, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.