Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEBPB Mouse anti-Human, Rat, Clone: 2B6E10, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | CEBPB |
|---|---|
| Clone | 2B6E10 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7067
![]()
|
Proteintech Group Inc
666491IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues.Specifications
| CEBPB | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| C/EBP beta, CEBPB, CRP2, IL6DBP, LAP, Liver activator protein, NF IL6, Nuclear factor NF IL6, TCF 5, TCF5, Transcription factor 5 | |
| CEBPB | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2B6E10 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P17676, P21272 | |
| 1051, 24253 | |
| CEBPB Fusion Protein Ag20073 | |
| Primary | |
| -20°C | |
| CEBPB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title