Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cecropin B Antibody, Novus Biologicals™
SDP

Catalog No. NB600867 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600867 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600867 Supplier Novus Biologicals Supplier No. NB600867
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cecropin B Polyclonal specifically detects Cecropin B in Silkworm samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Cecropin B
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Concentration 0.33 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA 1:5000-1:20000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 50% (v/v) Glycerol with 0.01% Sodium Azide
Gene Accession No. P04142
Gene Alias CECB1, CECB2, Lepidopteran A/B
Host Species Rabbit
Immunogen This affinity purified Cecropin B Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic full-length Cecropin B corresponding to a modified sequence from that which is found in silkworm: Lys-Trp-Lys-Val-Phe-Lys-Lys-Ile-Glu-Lys-Val-Gly-Arg-Asn-Ile-Arg-Asn-Gly-Ile-Val-Lys-Ala-Gly-Pro-Ala-Ile-Ala-Val-Leu-Gly-Glu-Ala-Lys-Ala-Leu. (Uniprot: P04142)
Molecular Weight of Antigen 6.83 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reactivity against other forms of cecropins has not yet been determined.
Target Species Silkworm
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.