Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefixime trihydrate

Catalog No. p-7057456
Click to view available options
Quantity:
100 mg
1 g

A broad spectrum third generation cephalosporin | CAS: 125110-14-7 | C16H15N5O7S2 | 453.44 g/mol

A broad spectrum third generation cephalosporin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125110-14-7
Molecular Formula C16H15N5O7S2
Molecular Weight (g/mol) 453.44
MDL Number MFCD00865020
InChI Key OKBVVJOGVLARMR-QSWIMTSFSA-N
PubChem CID 5362065
ChEBI CHEBI:472657
IUPAC Name (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-[(carboxymethoxy)imino]acetamido]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES NC1=NC(=CS1)C(=N\OCC(O)=O)\C(=O)N[C@H]1[C@H]2SCC(C=C)=C(N2C1=O)C(O)=O

Specifications

Type Cefixime Trihydrate
Physical Form Powder
Melting Point 218°C to 225°C
Color Yellow
Quantity 100 mg
Sensitivity Air and light sensitive
Solubility Information Soluble in water at 55;1 μg/mL
Formula Weight 507.49
Chemical Name or Material Cefixime trihydrate
Hazard Category H317-H334
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
RTECSNumber XI0367200
TSCA No
storageNote1 2° to 8°C
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.