Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEI Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CEI |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CEI Polyclonal specifically detects CEI in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
CEI | |
Polyclonal | |
Rabbit | |
Human | |
153571 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:HWGDDCYLTNRLWQDLKPPSHVENGQELRLAPPVQWALQVQGNQLQTAVLC | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
C5orf38, CEI IRX2NB, chromosome 5 open reading frame 38, coordinated expression to IRX2, coordinated expression to IRXA2 homeobox, Coordinated expression to IRXA2 protein, IRX2 neighbor, IRX2NB, MGC126657, MGC126683, protein CEI | |
C5ORF38 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title