Learn More
Invitrogen™ eBioscience™ Cell Stimulation Cocktail (plus protein transport inhibitors) (500X)
Catalog No. p-6933038
Encompass_Preferred
Click to view available options
Quantity:
100 μL
4 x 100 μL
Catalog No. 50-930-7
Supplier: Invitrogen™ 00497593
Only null left
Add to Cart
Add to Cart
Can be used for induction and subsequent intracellular detection of cytokines and other secreted proteins in both in vitro-cultured and ex vivo cells
Cocktail of phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A and monensin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines. Brefeldin A and Monensin result in the accumulation of secreted proteins in the endoplasmic reticulum and Golgi apparatus
  • A cocktail of phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A and monensin
  • Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines
  • Brefeldin A and Monensin result in the accumulation of secreted proteins in the endoplasmic reticulum and Golgi apparatus
  • Thus, it can be used for induction and subsequent intracellular detection of cytokines and other secreted proteins in both in vitro-cultured and ex vivo cells
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 μL
For Use With (Application) Flow Cytometry, Functional Assays, Cell Culture
Product Line eBioscience
Product Type Cell Stimulation Cocktail
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.