Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CellMask™ Orange Actin Tracking Stain

Catalog No. p-7191851
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 60 Assays
5 x 60 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A57244 1 x 60 Assays
A57247 5 x 60 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A57244 Supplier Invitrogen™ Supplier No. A57244
Only null left
Add to Cart
Edge
Add to Cart

Stain and track intracellular F-actin with CellMask Orange Actin Tracking Stain in live or fixed cells in ICC, IHC, and 3D cell culture. The stain easily permeates live-cell membranes, is retained within the cells after loading, and shows no detectable effects on viability after 24 hours of incubation.

Stain and track intracellular F-actin with CellMask Orange Actin Tracking Stain in live or fixed cells in immunofluorescence (IF), immunocytochemistry (ICC), and immunohistochemistry (IHC) protocols, and in 3D cell culture. The CellMask Orange Actin Tracking Stain fluorescently labels polymerized/filamentous actin (F-actin) in live or fixed cells because it is designed to readily permeate live cells, resulting in bright and uniform labeling. CellMask Orange Actin Tracking Stain can be used to track F-actin in live cells for 24 hours or more. Live cells stained with CellMask Orange Actin Tracking Stain can be fixed; the stain can also be used on already fixed cells. Cell Mask Orange Actin Tracking Stain can be multiplexed with antibodies in IF, ICC, and IHC protocols, and can also be used in 3D cell culture for 3D imaging.

Features of Cell Mask Orange Actin Tracking Stain include the following:
• Stains F-actin in live or fixed cells for fluorescence 2D monolayer or 3D imaging experiments
• Can be multiplexed with antibodies in IF/ICC/IHC experiments
• Fluorescence maxima (Excitation/Emission): 545/570 nm; can be detected with TRITC/RFP filter

CellMask actin tracking stains are available in three colors: green (Cat. No. A57243, A57246), detected with a FITC/GFP filter (Ex 503 nm/Em 512 nm), orange (Cat. No. A57244, A57247) detected with a TRITC/RFP filter (Ex 545 nm/Em 570 nm), and deep red (Cat. No. A57245, A57248), detected with a CY5/Deep Red filter (Ex 652 nm/Em 669 nm).

CellMask actin tracking stains detect F-actin in live or fixed cells or tissue with a targeting structure that closely resembles jasplakinolide. They do not label globular actin (G-actin) or monomer actin (M-actin). CellMask actin tracking stains easily permeate live cell membranes and are retained within live cells after loading. As demonstrated in the figures below, incubation with CellMask actin tracking stains has no detectable effect on viability of various cell lines after 24 hours of incubation. In addition, CellMask actin tracking stains can be used to detect F-Actin in paraformaldehyde fixed cells or tissue (freshly fixed or cryo frozen only) and can be multiplexed with antibody detection.

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Content And Storage Contains 1 vial with enough reagent to stain 120 slides, when 500 μL of 1X staining solution is used per slide.

• Add 60 μL/vial of anhydrous DMSO to make a 1000X (1 mM) stock solution
• Dissolve stock solution 1:1000 in cell imaging solution to make 1X (1 μM) staining solution

Store at room temperature. Can be stored to –20°C with no freeze/thaw cycles. When stored as instructed, these reagents are effective for a minimum of 6 months after receiving.

Excitation Wavelength Range 570 nm
Dye Type TRITC/RFP
For Use With (Equipment) Fluorescence Microscope, High Content Analyzer
Quantity 1 x 60 Assays
Detection Method Fluorescence Microscopy
Description CellMask™ Orange Actin Tracking Stain
Emission 545 nm
Product Line CellMask
Shipping Condition Room Temperature
Target F-Actin
Label Type Fluorescent Dye
Product Type Actin
SubCellular Localization Actin, Cytoskeleton
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.