Learn More
Thermo Scientific™ CellSensor™ c-Fos-bla HEK293T Cell Line
Description
This cell line can be used to detect agonists/antagonists of the c-fos pathway. The c-fos-bla HEK 293T cells have been shown to be responsive to phorbol 12-myristate 13-acetate (PMA) stimulation. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Quantity | 1 vial |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.