Learn More
Thermo Scientific™ CellSensor™ c-Fos-bla ME-180 Cell Line
Description
The CellSensor™ c-fos-bla ME-180 cell line contains a beta-lactamase reporter gene under control of the c-fos response element stably integrated into ME-180 cells. To construct this cell line, the c-fos-bla construct was transduced into ME-180 cells by lentivirus. Flow cytometry was used to isolate cells responsive to epidermal growth factor (EGF). This cell line is validated for DMSO tolerance, cell number, stimulation time, substrate loading time, as well as Z' and EC50 concentrations for EGF. The CellSensor™ c-fos-bla ME-180 cell line is responsive to EGF, HGF, IL-6, OSM, TNFα, IL-1-alpha; and PMA/Thaps, and can be used to probe the JNK/P38/MAPK and JAK/STAT signaling pathways. This cell line can be adapted for high-throughput screening of agonist or antagonist compound libraries. Candidate drugs can also be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Target Entry | STAT1, STAT3, JAK1, EGFR, ERK1/2, p38, MAPK pathway, RAF, JAK2, c-MET, JNK, JAK/STAT (IFN gamma, IL-6/OSM) pathway, RON, MEK |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.