Learn More
Thermo Scientific™ CellSensor™ CRE-bla HEK 293T Cell Line
Description
The CellSensor™ CRE-bla HEK 293T cell line contains the b-lactamase reporter gene under control of the cAMP response element (CRE). To obtain the cell line, the CRE-bla construct was transduced into HEK 293T cells by lentivirus. Subsequent flow cytometry was used to isolate clones responsive to forskolin stimulation. The CellSensor™ CRE-bla HEK 293T cell line can be used to build specific G-protein coupled receptor (GPCR) assays or to detect changes in intracellular cAMP levels. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Target Entry | PKA, cAMP, cAMP/PKA pathway |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.