Learn More
Thermo Scientific™ CellSensor™ DHFR/E2F-bla NIH3T3 Cell Line
Description
The CellSensor™ dhfr(E2F)-bla NIH 3T3 Cell Line contains a beta-lactamase reporter gene under control of the E2F/DP1 binding sequence from the DHFR gene promoter, stably integrated into NIH 3T3 cells. This cell line is a clonal population isolated by flow cytometry in response to 10% newborn bovine serum. This cell line has been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time, and validated for Z-factor using newborn bovine serum. Additional data using alternate stimuli are shown. The G1/S cell-cycle checkpoint controls the passage of eukaryotic cells from the first gap phase (G1) into the DNA synthesis phase (S). Two cell-cycle kinases, CDK4/6-cyclin D and CDK2-cyclin E, and the transcription complex that includes Rb and E2F, are pivotal in controlling this checkpoint. During G1, the Rb-HDAC repressor complex binds to the E2F-DP1 transcription factors, inhibiting downstream transcription. Phosphorylation of Rb by CDK4/6 and CDK2 dissociates the Rb-repressor complex, permitting transcription of S-phase genes encoding for proteins that amplify the G1-to-S switch and that are required for DNA replication. Many different stimuli exert checkpoint control, including TGFβ, DNA damage, contact inhibition, replicative senescence, and growth factor withdrawal. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Quantity | 1 Vial |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.