Learn More
Thermo Scientific™ CellSensor™ GAS-bla ME-180 Cell Line
Description
To construct this cell line, the GAS-bla construct was transduced into ME-180 cells by lentivirus. Flow cytometry was used to isolate cells responsive to Interferon Gamma (IFN-γ). This cell line is validated for DMSO tolerance, cell number, stimulation time, substrate loading time, as well as Z'and EC50 concentrations for IFN-γ. The CellSensor™ GAS-bla ME-180 cell line is responsive to IFN-γ and can be used to probe the JAK/STAT signaling pathway. This cell line can be adapted for high-throughput screening of agonist or antagonist compound libraries. Candidate drugs can be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Quantity | 1 vial |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.