Learn More
Invitrogen™ CellSensor™ LEF/TCF-bla SW480 Cell Line
Description
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Product Line | CellSensor |
| Quantity | 1 vial |
| Target Entry | Beta-Catenin, Wnt/Beta-Catenin pathway |
| Type | Cell Line |
| Cell Line | SW480 |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.