Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CellSensor™ NFAT-bla CHO-K1 Cell Line

Catalog No. K1534
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1534 1 vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1534 Supplier Invitrogen™ Supplier No. K1534
Only null left
Add to Cart
Add to Cart
Request a Quote

CellSensor™ NFAT-bla CHO-K1 Cell Line

The G protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPCRs are instrumental in the transmission of a wide range of chemical messages from the extracellular environment to the interior of the cell. GPCRs are involved in a wide range of disorders, including allergies, cardiovascular dysfunction, depression, obesity, cancer, pain, diabetes, and various central nervous system disorders. GPCR signaling is mediated by trimeric G-proteins containing α, β, and γ subunits and can be categorized into signaling classes based upon subunit composition. G alpha, G alpha s, and G alpha i/o proteins mediate intracellular signaling through activation of signaling pathways leading to distinct physiological endpoints. Activation of G alpha s and G alpha i/o coupled receptors leads to stimulation or inhibition of adenylate cyclase, respectively; activation of G alpha q coupled receptors results in stimulation of phospholipase C. GPCR signaling through these distinct pathways can be monitored by activation of specific transcriptional response elements placed upstream of a reporter gene. Invitrogen has developed GeneBLAzer™ Master Cell Lines employing the beta-lactamase reporter system, which can be used to screen for both agonists and antagonists of GPCRs coupling to each of the above signaling pathways. These cell-based assays make use of a membrane permeant fluorescent substrate (CCF4-AM or CCF2-AM), enabling a ratiometric readout that reduces errors due to sample variability, leads to excellent Z'-factor values, and allows for assay miniaturization. GeneBLAzer™ Master Cell Lines with either NFAT or CRE response elements are available in Jurkat, CHO-K1, or FreeStyle™ 293F cell backgrounds (six cell lines in total) for the development of assays in suspension or adherent cell format. Stable cell lines employing an NFAT response element upstream of the beta-lactamase gene are designed for the study of Gq coupled receptors. Stable cell lines employing the cAMP response element (CRE) upstream of the beta-lactamase gene are designed for the study of Gs coupled receptors. FreeStyle™ 293F cells are HEK 293 cells that have been adapted for growth in suspension in serum-free media. Alternatively, these cells may be grown in the presence of serum for use in the adherent format. Academic and non-profit customers, please inquire for special pricing.
TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Product Line CellSensor
Quantity 1 vial
Target Entry PKC, PKC and calcium pathway
Type CHO-K1 Cells
Cell Line CHO-K1
System Type CellSensor™
Druggable Target GPCR

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.