Learn More
Thermo Scientific™ CellSensor™ NFAT-bla Jurkat Cell Line
Description
The CellSensor™ NFAT-bla Jurkat cell line contains a beta-lactamase reporter gene under control of a Nuclear Factor of Activated T Cells (NFAT) response element stably integrated into Jurkat cells. This cell line can be used as a parental cell line to build specific G-protein coupled receptor (GPCR) assays after transfection of additional genes of interest. NFAT-bla Jurkat cells have been shown to be responsive to phorbol 12-myristate 13-acetate (PMA) and Ionomycin stimulation. Additional testing data using alternate stimuli is available upon request. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Quantity | 1 vial |
| Type | Cell Line |
| Cell Line | Jurkat |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.