Learn More
Thermo Scientific™ CellSensor™ NFkB-bla RAW 264.7 Cell Line
Description
The CellSensor™ NFκB-bla RAW264.7 mouse macrophage cell line contains a beta-lactamase reporter gene under control of a stably integrated NFκB response element. This cell line is a clonal population isolated in response to lipopolysaccharide by flow cytometry. This cell line has been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time, and has been validated for Z'-factor and EC50 concentrations of the TLR7 ligand imiquimod. Additional data using alternate stimuli are shown. The molecules responsible for coordinating the immune systems recognition of pathogens are Toll-like receptors (TLRs), of which there are currently 10 identified members in humans. TLRs are pattern recognition receptors (PRRs), which bind to pathogen-associated molecular patterns (PAMPs), small molecular sequences consistently found on pathogens. This binding activates the transcription of immune system genes and regulators such as cytokines, chemokines, and costimulatory molecules, thereby promoting the initial immune response of macrophages and neutrophils. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Target Entry | TNF alpha, NIK, TLR, I kappa B, IRAK, IKK, NF-kappa B pathway |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.