Learn More
Thermo Scientific™ CellSensor™ p53RE-bla HCT-116 Cell Line
Description
The CellSensor™ p53RE-bla HCT-116 cell line contains a beta-lactamase reporter gene under control of the p53 response element stably integrated into HCT-116 cells. To construct this cell line, the p53RE-bla construct was transduced into HCT-116 cells by lentivirus. Flow cytometry was used to isolate cells responsive to Mitomycin. This cell line was validated for DMSO tolerance, cell number, stimulation time, substrate loading time, as well as Z'; and EC50 concentrations for Mitomycin. The CellSensor™ p53RE-bla HCT-116 cell line is responsive to Mitomycin and can be used to probe p53 signaling, which is involved in DNA repair, cell cycle arrest, and in some cases, apoptosis. This cell line can be adapted for high-throughput screening for agonists or antagonists with compound libraries. Candidate drugs can be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Target Entry | p53, DNA damage/p53 response pathway |
| Type | Cell Line |
| Cell Line | HCT-116 |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.