Learn More
Thermo Scientific™ CellSensor™ TNFa-bla THP-1 Cell Line
Description
Tumor necrosis factor-alpha (TNFα) cytokine cell signaling is a critical pathway involved in immune and inflammatory responses. In addition, TNFα has been shown to inhibit growth and induce differentiation in tumor and non-tumor cells. The CellSensor™ TNFα-bla THP-1 cell line is a suspension cell line derived from human acute monocytic leukemia cells. The TNFα-bla THP-1 cell line is used to measure the TNFα cytokine signaling pathway via transcriptional regulation of the beta-lactamase gene. This cell line was constructed using GenomeScreen™ technology utilizing a promoterless beta-lactamase reporter gene randomly integrated into the genome of the THP-1 cell line. Sequential rounds of sorting were performed to isolate cell clones containing tagged genes induced by TNFα. These cells have been shown to be responsive to TNFa agonist. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Quantity | 1 vial |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.