Learn More
Invitrogen™ CellSensor™ TrkB-NFAT-bla CHO-K1 Cell Line
Description
BDNF (brain-derived neurotrophic factor) and its receptor TrkB (also known as NTRK2) play a fundamental role in regulating neural development, survival, and synaptic activity and plasticity. TrkB is also a potential anticancer target, since altered signaling through TrkB promotes tumor formation, survival, and metastasis of various cancers (including neuroblastomas, multiple myelomas, and pancreatic ductal adenocarcinomas). Moreover, growing evidence indicates that BDNF and its receptor influences food intake and body weight control.
CellSensor™ TrkB-NFAT-bla CHO-K1 cells contain a beta-lactamase reporter gene under control of the NFAT Response Element that has been stably integrated into CHO-K1 cells along with TrkB. CellSensor™ TrkB-NFAT-bla CHO-K1 cells express beta-lactamase upon stimulation with brain-derived neurotrophic factor (BDNF). This cell line is a clonal population isolated by flow cytometry and has been tested for robust performance by assessing a variety of assay parameters.
Specifications
Specifications
| Content And Storage | CellSensor™ TrkB-NFAT-bla CHO-K1 cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Target Entry | TRKB, PKC and calcium pathway |
| Quantity | 1 vial |
| Product Type | TrkB-NFAT-bla CHO-K1 Cell Line |
| Product Line | CellSensor |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.