Learn More
Thermo Scientific™ CellSensor™ TrkC-NFAT-bla CHO-K1 Cell Line
Description
Neurotrophins (NGF, BDNF, NT-3, and NT-4) and their transmembrane receptors (TrkA, TrkB, TrkC, and P75NTR) play important roles in the regulation of neuronal and non-neuronal cell proliferation, differentiation, survival, and death. Neurotrophin signaling also mediates higher-order neuronal activities, such as learning, memory, and behavior. Alterations in neurotrophin levels and their receptors have been implicated in neurodegenerative disorders such as Alzheimer's disease and Huntington's disease, as well as psychiatric disorders. Neurotrophins propagate their signal through activating multiple signaling pathways. One of the signaling pathways of NT-3 (the ligand for TrkC) activates phospholipase C, releasing DAG and IP3, increasing downstream intracellular calcium, and activating protein kinase C. This in turn promotes the translocation of the transcription factor-nuclear factor of activated T-cells (NFAT)-from the cytosol into the nucleus, resulting in NFAT-dependent transcription.
The CellSensor™ TrkC-NFAT-bla CHO-K1 Cell Line was engineered by integrating the human TrkC expression plasmid into the genome of the CellSensor™ NFAT-bla CHO-K1 cell line, which is engineered to express beta-lactamase under the control of NFAT. This cell line has been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, substrate loading time, and in cryopreserved cells, and has been validated for Z'-factor and EC50 values under optimized conditions using NT-3. Additional testing information using various small-molecule inhibitors and RNAi has been performed.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ TrkC-NFAT-bla CHO-K1 cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Detection Method | Fluorescence |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Target Entry | PKC and calcium pathway, TRKC |
| Type | CHO-K1 Cells |
| Cell Line | CHO-K1 |
| System Type | CellSensor™ |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.