Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellulose microcrystalline, extra pure, average particle size 90μm, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
25kg
Packaging:
Glass bottle
Plastic bag in resealable cardbox
Plastic drum
9004-34-6
9004-34-6
Filter aid
Linear Formula (C12H20O10)n
Heavy Metals 10ppm max.
Solubility Information Solubility in water: insoluble.
Physical Form Fine Crystalline Powder or Granular Powder
Infrared Spectrum Authentic
Grade Extra Pure
Loss on Drying 1.5% max.
Particle Size 250 Micron <1% min., 75 Micron >45% min., 32 Micron >70% min.
Packaging Glass bottle
Residue after Ignition 0.05% max.
Solubility Solubility in water: insoluble.
Color White
pH 5 to 7
Quantity 250g
Chemical Name or Material Cellulose microcrystalline
Show More Show Less