Learn More
Invitrogen™ CellVue Diluent C (For CellVue™ Cell Labeling Kits)
Click to view available options
Quantity:
10 mL
Description
Diluent C is the Buffer required for the dilution of the dyes in the CellVue™ Cell Labeling Kits. Please refer to the specific labeling kit for instructions of use.
Reported Application
Flow Cytometric Analysis, Microscopy
Reported Application
Flow Cytometric Analysis, Microscopy
Specifications
Specifications
Quantity | 10 mL |
Validated Application | Cell Analysis, Flow Cytometry |
Form | Kit |
Reagent Type | Diluent |
Product Type | Cell Labeling |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.