Learn More
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Detection Method | Microscopy |
| For Use With (Application) | Fluorimetry |
| Includes | Human Killer Cell Base Media 1, Recombinant Human IFN-γ, Recombinant Human IL-2, Mouse Anti-Human CD3, Reconstitution Buffer 1, Reconstitution Buffer 2 |
| Quantity | 1 Kit |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
