Learn More
R&D Systems™ CellXVivo Human M1 Macrophage Differentiation Kit
SDP
Catalog No. CDK012 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
CDK012 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. CDK012 Supplier R&D Systems™ Supplier No. CDK012
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
For the differentiation of human CD14+ monocytes into M1 macrophages
Specifications
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
For Use With (Application) Cell Differentiation
Product Line R&D Systems
Quantity 1 Kit