Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CellXVivo™ Human Th2 Cell Differentiation Kit
SDP

Catalog No. CDK002 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDK002 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. CDK002 Supplier R&D Systems™ Supplier No. CDK002

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the differentiation of Th2 cells from a preparation of naïve CD4+ T cells.

Specifications

Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Includes Mouse Anti-Human CD3 Antibody
Human Th2 Differentiation Supplements
Reconstitution Buffers
Wash Buffer (20X)
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.