Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Includes | Mouse Anti-Human CD3 Antibody Human Treg Reagent 1 Human Treg Reagent 2 Human Treg Reagent 3 Human Treg Reagent 4 Reconstitution Buffer 1 Reconstitution Buffer 2 Wash Buffer (20X) |
| Product Line | R&D Systems |
| Quantity | 1 Kit |