Learn More
Description
Specifications
Specifications
| Antigen | CELSR2 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 594 |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | cadherin EGF LAG seven-pass G-type receptor 2, Cadherin family member 10, cadherin, EGF LAG seven-pass G-type receptor 2 (flamingo homolog, Drosophila), CDHF10FLJ34118, EGFL2FLJ45143, EGF-like protein 2, EGF-like-domain, multiple 2, epidermal growth factor-like 2, Epidermal growth factor-like protein 2, Flamingo homolog 3, Flamingo1, FLJ45845, KIAA0279FLJ42737, MEGF3cadherin, EGF LAG seven-pass G-type receptor 2, flamingo (Drosophila) homolog, Multiple EGF-like domains protein 3, multiple epidermal growth factor-like domains 3, Multiple epidermal growth factor-like domains protein 3 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human CELSR2 fragment. |
| Purification Method | Antigen Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
