Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CeMMEC1
GREENER_CHOICE

Catalog No. p-7237064
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

TAF1 inhibitor

CeMMEC1 is a TAF1 inhibitor (Kd = 1.8 μM, IC50 = 900 nM). CeMMEC1 impairs cell viability in combination with (+)-JQ1 (Cat. No. 4499) and inhibits proliferation of THP-1 and H23 lung adenocarcinoma cells.

Specifications

Chemical Name or Material N-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1,2-dihydro-2-methyl-1-oxo-4-isoquinolinecarboxamide
CAS 440662-09-9
Quantity 10 mg
Target Bromodomain Inhibitors
Molecular Formula C19H16N2O4
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.