Learn More
Bullard™ CEN10™ Sleek Low Profile Helmet
Bullard CEN10™ Sleek Low Profile Helmet features sleek, low-profile vented and non-vented designs, self-adjusting patented FlexGlide™ suspension system, an impact resistant helmet shell with inner foam comfort liner, and an integrated 3-point chinstrap.
$121.98 - $122.10
Specifications
Dimensions (L x W x H) | 11 x 8.75 x 6.25 in. |
---|---|
Liner Material | EPS Foam |
Material | PC ABS Blend |
Head Style | FlexGuide Head Suspension System |
Catalog Number | Mfr. No. | Ventilated | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Ventilated | Color | Price | Quantity & Availability | ||||
19-160-570
![]() |
Bullard™
C10VYLAMR |
Vented | Yellow |
Each for $122.10
Case of 5 EA for $583.00
|
|
||||
19-160-560
![]() |
Bullard™
C10NWHAMR |
Not Vented | White |
Each for $122.10
Case of 5 EA for $583.00
|
|
||||
19-160-561
![]() |
Bullard™
C10NRDAMR |
Not Vented | Red |
Each for $122.10
Case of 5 EA for $583.00
|
|
||||
19-160-562
![]() |
Bullard™
C10NDGAMR |
Not Vented | Dove Gray |
Each for $122.10
Case of 5 EA for $583.00
|
|
||||
19-160-563
![]() |
Bullard™
C10NYLAMR |
Not Vented | Yellow |
Each for $122.10
Case of 5 EA for $583.00
|
|
||||
19-160-564
![]() |
Bullard™
C10NHYAMR |
Not Vented | Hi-vis Yellow |
Each for $122.10
Case of 5 EA for $583.00
|
|
||||
19-160-565
![]() |
Bullard™
C10NORAMR |
Not Vented | Orange |
Each for $122.10
Case of 5 EA for $583.00
|
|
||||
19-160-567
![]() |
Bullard™
C10VWHAMR |
Vented | White |
Each for $122.10
Case of 5 EA for $583.00
|
|
||||
19-160-568
![]() |
Bullard™
C10VRDAMR |
Vented | Red |
Each for $121.98
Case of 5 EA for $602.41
|
|
||||
19-160-569
![]() |
Bullard™
C10VDGAMR |
Vented | Dove Gray |
Each for $122.10
Case of 5 EA for $583.00
|
|
||||
19-160-571
![]() |
Bullard™
C10VHYAMR |
Vented | Hi-vis Yellow |
Each for $122.10
Case of 5 EA for $583.00
|
|
||||
19-160-572
![]() |
Bullard™
C10VORAMR |
Vented | Orange |
Each for $122.10
Case of 5 EA for $583.00
|
|
||||
Description
- The patented FlexGlide suspension system lets wearers auto-adjust the fit of their helmet for all day comfort
- The CEN10 is the only auto-adjusting suspension helmet on the market, allowing even the smallest adjustments to achieve the perfect fit
- Replaceable brow pad, comfort liner, and nape cover for ultimate utility
- Distinctive three rib trademark
- Manufacturer warranty: 3 years
- Life span: 5 years
Specifications
11 x 8.75 x 6.25 in. | |
EPS Foam | |
PC ABS Blend | |
FlexGuide Head Suspension System |