Learn More
Cenexin-1 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $649.44
Specifications
| Antigen | Cenexin-1 |
|---|---|
| Concentration | 1.53 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0908
![]()
|
Rockland Immunochemicals
200401C43 |
100 μg |
Each for $649.44
|
|
|||||
50-199-0909
![]()
|
Rockland Immunochemicals
200401C43S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. It is directed against, and shows specific reactivity for, human Cenexin-1 protein. A BLAST analysis was used to suggest cross-reactivity with Cenexin-1 from human, chimpanzee, mouse, rat, bovine sources based on 100% homology with the immunizing sequence. Reactivity with Cenexin-1 protein from other sources has not been determined.
Seems to be a major component of sperm tail outer dense fibers (ODF). ODFs are filamentous structures located on the outside of the axoneme in the midpiece and principal piece of the mammalian sperm tail and may help to maintain the passive elastic structures and elastic recoil of the sperm tail. May have a modulating influence on sperm motility. Functions as a general scaffold protein that is specifically localized at the distal/subdistal appendages of mother centrioles. Component of the centrosome matrix required for the localization of PLK1 and NIN to the centrosomes. Required for the formation and/or maintenance of normal CETN1 assembly.Specifications
| Cenexin-1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q5BJF6 | |
| 4957 | |
| This protein-A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a truncated recombinant protein hCenexin1. | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| ODF2 |
| 1.53 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| 84 kDa outer dense fiber protein; AI848335; cancer/testis antigen 134; Cenexin; cenexin 1; CT134; KKT4; MMTEST29; ODF2; ODF2/1; ODF2/2; Odf84; outer dense fiber of sperm tail 2; outer dense fiber of sperm tails 2; Outer dense fiber of sperm tails protein 2; outer dense fiber of sperm tails, 84-kD; Outer dense fiber protein 2; RP11-187B3.1; sperm outer dense fiber major protein 2; sperm tail structural protein; testis tissue sperm-binding protein Li 51e; testis-specific autoantigen | |
| ODF2 | |
| Primary | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.