Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPB Antibody, Novus Biologicals™
SDP

Catalog No. NB1002369 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002369 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002369 Supplier Novus Biologicals Supplier No. NB1002369
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CENPB Polyclonal specifically detects CENPB in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CENPB
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:100-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. P07199
Gene Alias CENP-B, centromere autoantigen B, Centromere protein B, centromere protein B (80kD), centromere protein B, 80kDa, major centromere autoantigen B
Gene Symbols CENPB
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 540 and the C-terminus (residue 599) of human Centromere Protein B using the numbering given in entry SwissProt entry P07199 (GeneID 1059).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1059
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.