Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPF Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB500101SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500101SS 0.025 mg
NB500101 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500101SS Supplier Novus Biologicals Supplier No. NB500101SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 49 publications

CENPF Polyclonal specifically detects CENPF in Human, Mouse, Bovine, Equine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount, Knockdown Validated.

Specifications

Antigen CENPF
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1500-1:2500, Flow Cytometry 1:50-1:200, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200-1:1000immunoprecipitation 1:50, Immunoprecipitation 1:50, Immunohistochemistry-Paraffin reported in scientific literature (PMID 20398247), Immunohistochemistry-Frozen reported in scientific literature (PMID 26195156), Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 24876181), KnockDown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P49454
Gene Alias AH antigen, cell-cycle-dependent 350K nuclear protein, CENF, CENP-F, CENP-F kinetochore protein, centromere protein F, centromere protein F (350/400kD, mitosin), centromere protein F, 350/400ka (mitosin), centromere protein F, 350/400kDa (mitosin), hcp-1, Kinetochore protein CENPF, mitosin, PRO1779
Gene Symbols CENPF
Host Species Rabbit
Immunogen A bacterial fusion protein from the C-terminus of human CENPF. [UniProt# P49454]
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 1063
Target Species Human, Bovine
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.