Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPF, Rabbit anti-Human,Mouse,Bovine,Equine, DyLight 650, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB004065 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB004065 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB004065 Supplier Novus Biologicals Supplier No. NB500101C0.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CENPF Polyclonal specifically detects CENPF in Human, Mouse, Bovine, Equine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount, Knockdown Validated.

Specifications

Antigen CENPF
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount
Formulation 50 mM Sodium Borate
Gene Alias AH antigen, cell-cycle-dependent 350K nuclear protein, CENF, CENP-F, CENP-F kinetochore protein, centromere protein F, centromere protein F (350/400kD, mitosin), centromere protein F, 350/400ka (mitosin), centromere protein F, 350/400kDa (mitosin), hcp-1, Kinetochore protein CENPF, mitosin, PRO1779
Gene Symbols CENPF
Host Species Rabbit
Immunogen A bacterial fusion protein from the C-terminus of human CENPF. [UniProt# P49454]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 1063
Target Species Human, Mouse, Bovine, Equine
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.