Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPL Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB032578 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB032578 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB032578 Supplier Novus Biologicals Supplier No. NBP2846500.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CENPL Polyclonal specifically detects CENPL in Human samples. It is validated for Western Blot.

Specifications

Antigen CENPL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias C1orf155, CENP-L, centromere protein L, chromosome 1 open reading frame 155, dJ383J4.3, FLJ31044, FLJ31786, ICEN33, Interphase centromere complex protein 33, RP3-383J4.1
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of Human CENPL
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91687
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.