Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPO Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB358663 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358663 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358663 Supplier Novus Biologicals Supplier No. NBP176240MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CENPO Polyclonal antibody specifically detects CENPO in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen CENPO
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias CENP-OMGC11266, centromere protein O, centromeric protein O, ICEN36, ICEN-36, Interphase centromere complex protein 36, MCM21R
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the center of human CENPO. The immunogen is located within amino acids 170 - 220 of CENPO. Amino Acid Squence: AYSGRKYQADRLQSDF
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79172
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.