Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPQ Antibody (OTI1C5), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N271991A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271991A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271991A700 Supplier Novus Biologicals Supplier No. NBP271991AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CENPQ Monoclonal specifically detects CENPQ in Human samples. It is validated for Western Blot.

Specifications

Antigen CENPQ
Applications Western Blot
Classification Monoclonal
Clone OTI1C5
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C6orf139, CENP-QFLJ10545, centromere protein Q, chromosome 6 open reading frame 139
Gene Symbols CENPQ
Host Species Mouse
Immunogen Full length recombinant protein of human CENPQ (NP_060602) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 55166.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.