Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPQ Antibody (OTI1C5), FITC, Novus Biologicals™
SDP

Catalog No. NB288461 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288461 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288461 Supplier Novus Biologicals Supplier No. NBP271991F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CENPQ Monoclonal antibody specifically detects CENPQ in Human samples. It is validated for Western Blot

Specifications

Antigen CENPQ
Applications Western Blot
Classification Monoclonal
Clone OTI1C5
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias C6orf139, CENP-QFLJ10545, centromere protein Q, chromosome 6 open reading frame 139
Host Species Mouse
Immunogen Full length human recombinant protein of human CENPQ (NP_060602) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 55166
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.