Learn More
Invitrogen™ CENPT Polyclonal Antibody

Description
Predicted to react with Rat samples.
Specifications
Specifications
| Antigen | CENPT |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | CENPT |
| Gene Accession No. | Q561R1, Q96BT3 |
| Gene Alias | C16orf56; CENPT; CENP-T; Centromere protein T; G630055P03Rik; ICEN22; Interphase centromere complex protein 22; RGD1309383 |
| Gene Symbols | CENPT |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.