Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Centaurin alpha 1 Antibody (3E2D9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261732
Description
Centaurin alpha 1 Monoclonal specifically detects Centaurin alpha 1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Centaurin alpha 1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
arf-GAP with dual PH domain-containing protein 1, ArfGAP with dual PH domains 1, CENTA1centaurin-alpha-1, centaurin, alpha 1, centaurin-alpha, Centaurin-alpha-1, Cnt-a1, GCS1L, p42IP4, Putative MAPK-activating protein PM25 | |
Mouse | |
43.4 kDa | |
0.1 mL | |
Primary | |
Human, Rat | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
3E2D9 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
ADAP1 | |
Purified recombinant fragment of human ADAP1 (AA: 240-370) expressed in E. Coli. | |
Protein G purified | |
Signal Transduction | |
11033 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction