Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Centaurin alpha 1 Antibody (3E2D9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | Centaurin alpha 1 |
---|---|
Clone | 3E2D9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61732
![]() |
Novus Biologicals
NBP261732 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
Centaurin alpha 1 Monoclonal specifically detects Centaurin alpha 1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Centaurin alpha 1 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
Human, Rat | |
11033 | |
Purified recombinant fragment of human ADAP1 (AA: 240-370) expressed in E. Coli. | |
Primary | |
43.4 kDa |
3E2D9 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Signal Transduction | |
arf-GAP with dual PH domain-containing protein 1, ArfGAP with dual PH domains 1, CENTA1centaurin-alpha-1, centaurin, alpha 1, centaurin-alpha, Centaurin-alpha-1, Cnt-a1, GCS1L, p42IP4, Putative MAPK-activating protein PM25 | |
ADAP1 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title